Learn More
Biomatik Corporation ADGRL4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509368
Encompass_Preferred
The ADGRL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Adhesion G protein-coupled receptor L4 protein (85-299AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Adhesion G protein-coupled receptor L41 Publication, ADGRL41, EGF, latrophilin and seven transmembrane domain-containing protein 1, EGF-TM7-latrophilin-related protein, ETL protein. UniProt: Q9HBW9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9368 Supplier Biomatik Corporation Supplier No. CAC09786
The ADGRL4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Adhesion G protein-coupled receptor L4 protein (85-299AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Adhesion G protein-coupled receptor L41 Publication, ADGRL41, EGF, latrophilin and seven transmembrane domain-containing protein 1, EGF-TM7-latrophilin-related protein, ETL protein. UniProt: Q9HBW9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.