Learn More
Biomatik Corporation ZDHHC4 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509371
Encompass_Preferred
Zdhhc4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZDHHC4. The Zdhhc4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Probable Palmitoyltransferase Zdhhc4 Protein (277-344Aa). Applications: ELISA, WB, IHC. Target Synonyms: ZDHHC4. Accession Number: Q9NPG8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9371 Supplier Biomatik Corporation Supplier No. CAC09789
Zdhhc4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZDHHC4. The Zdhhc4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Probable Palmitoyltransferase Zdhhc4 Protein (277-344Aa). Applications: ELISA, WB, IHC. Target Synonyms: ZDHHC4. Accession Number: Q9NPG8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.