Learn More
Biomatik Corporation FAM105A anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09793
Encompass_Preferred
The FAM105A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Inactive ubiquitin thioesterase FAM105A protein (81-356AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Inactive ubiquitin thioesterase FAM105A, FAM105A. UniProt: Q9NUU6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9375