Learn More
Biomatik Corporation DNAJC17 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509376
Encompass_Preferred
The DNAJC17 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DnaJ homolog subfamily C member 17 protein (1-304AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: DnaJ homolog subfamily C member 17, DNAJC17. UniProt: Q9NVM6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9376 Supplier Biomatik Corporation Supplier No. CAC09794
The DNAJC17 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human DnaJ homolog subfamily C member 17 protein (1-304AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: DnaJ homolog subfamily C member 17, DNAJC17. UniProt: Q9NVM6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.