Learn More
Biomatik Corporation SLC6A15 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509393
Encompass_Preferred
Slc6A15 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC6A15. The Slc6A15 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sodium-Dependent Neutral Amino Acid Transporter B (0) At2 Protein (357-452Aa). Applications: ELISA, IHC, IF. Target Synonyms: SLC6A15. Accession Number: Q9H2J7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9393 Supplier Biomatik Corporation Supplier No. CAC09811
Slc6A15 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC6A15. The Slc6A15 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sodium-Dependent Neutral Amino Acid Transporter B (0) At2 Protein (357-452Aa). Applications: ELISA, IHC, IF. Target Synonyms: SLC6A15. Accession Number: Q9H2J7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.