Learn More
Biomatik Corporation LDAH anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509395
Encompass_Preferred
Ldah Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LDAH. The Ldah Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lipid Droplet-Associated Hydrolase Protein (1-325Aa). Applications: ELISA, WB, IHC. Target Synonyms: LDAH. Accession Number: Q9H6V9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9395 Supplier Biomatik Corporation Supplier No. CAC09813
Ldah Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LDAH. The Ldah Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Lipid Droplet-Associated Hydrolase Protein (1-325Aa). Applications: ELISA, WB, IHC. Target Synonyms: LDAH. Accession Number: Q9H6V9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.