Learn More
Biomatik Corporation MAP10 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09834
Encompass_Preferred
Map10 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MAP10. The Map10 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Microtubule-Associated Protein 10 Protein (401-600Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: MAP10. Accession Number: Q9P2G4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9416