Learn More
Biomatik Corporation COPS7A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09835
Encompass_Preferred
The COPS7A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human COP9 signalosome complex subunit 7a protein (2-275AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200. Alternative Names: SGN7a, Signalosome subunit 7a, Dermal papilla-derived protein 10, JAB1-containing signalosome subunit 7a, COPS7A, CSN7A, DERP10. UniProt: Q9UBW8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9417