Learn More
Biomatik Corporation SLCO1B3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09854
Encompass_Preferred
Slco1B3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLCO1B3. The Slco1B3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Solute Carrier Organic Anion Transporter Family Member 1B3 Protein (648-695Aa). Applications: ELISA, IHC, IF. Target Synonyms: SLCO1B3. Accession Number: Q9NPD5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9436