Learn More
Biomatik Corporation ZDHHC3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09862
Encompass_Preferred
Zdhhc3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to ZDHHC3. The Zdhhc3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Palmitoyltransferase Zdhhc3 Protein (94-171Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: ZDHHC3. Accession Number: Q9NYG2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9444