Learn More
Biomatik Corporation CRISPLD2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09873
Encompass_Preferred
The CRISPLD2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Cysteine-rich secretory protein LCCL domain-containing 2 protein (40-305AA). Applications: ELISA, WB. Recommended dilution: WB:1:500-1000. Alternative Names: Cysteine-rich secretory protein LCCL domain-containing 2, Cysteine-rich secretory protein 11, CRISP-11, LCCL domain-containing cysteine-rich secretory protein 2, CRISPLD2, CRISP11, LCRISP2. UniProt: Q9H0B8. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9455