Learn More
Biomatik Corporation RSPO3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509485
Encompass_Preferred
Rspo3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RSPO3. The Rspo3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human R-Spondin-3 Protein (22-272Aa). Applications: ELISA, IHC. Target Synonyms: RSPO3. Accession Number: Q9BXY4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9485 Supplier Biomatik Corporation Supplier No. CAC09903
Rspo3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RSPO3. The Rspo3 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human R-Spondin-3 Protein (22-272Aa). Applications: ELISA, IHC. Target Synonyms: RSPO3. Accession Number: Q9BXY4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.