Learn More
Biomatik Corporation NANOG anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09911
Encompass_Preferred
Nanog Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NANOG. The Nanog Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Homeobox Protein Nanog Protein (1-305Aa). Applications: ELISA, WB, IHC. Target Synonyms: NANOG. Accession Number: Q9H9S0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9493