Learn More
Biomatik Corporation LSM7 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09926
Encompass_Preferred
Lsm7 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to LSM7. The Lsm7 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human U6 Snrna-Associated Sm-Like Protein Lsm7 Protein (2-103Aa). Applications: ELISA, WB, IHC. Target Synonyms: LSM7. Accession Number: Q9UK45. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9508