Learn More
Biomatik Corporation TBC1D24 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509511
Encompass_Preferred
Tbc1D24 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TBC1D24. The Tbc1D24 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tbc1 Domain Family Member 24 Protein (1-169Aa). Applications: ELISA, IHC, IF. Target Synonyms: TBC1D24. Accession Number: Q9ULP9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9511 Supplier Biomatik Corporation Supplier No. CAC09929
Tbc1D24 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TBC1D24. The Tbc1D24 Polyclonal Antibody has been validated for the following applications: ELISA, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tbc1 Domain Family Member 24 Protein (1-169Aa). Applications: ELISA, IHC, IF. Target Synonyms: TBC1D24. Accession Number: Q9ULP9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.