Learn More
Biomatik Corporation ANGPTL2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509517
Encompass_Preferred
The ANGPTL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Angiopoietin-related protein 2 protein (23-493AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Angiopoietin-like protein 2, UNQ170/PRO196, ARP2, ANGPTL2. UniProt: Q9UKU9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9517 Supplier Biomatik Corporation Supplier No. CAC09935
The ANGPTL2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Angiopoietin-related protein 2 protein (23-493AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: Angiopoietin-like protein 2, UNQ170/PRO196, ARP2, ANGPTL2. UniProt: Q9UKU9. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.