Learn More
Biomatik Corporation GPR34 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09940
Encompass_Preferred
The GPR34 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Probable G-protein coupled receptor 34 protein (1-61AA). Applications: ELISA, WB, IF. Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200. Alternative Names: Probable G-protein coupled receptor 34, GPR34. UniProt: Q9UPC5. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9522