Learn More
Biomatik Corporation SLC13A4 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09944
Encompass_Preferred
Slc13A4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SLC13A4. The Slc13A4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Solute Carrier Family 13 Member 4 Protein (134-273Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: SLC13A4. Accession Number: Q9UKG4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9526