Learn More
Biomatik Corporation MINPP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509528
Encompass_Preferred
Minpp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MINPP1. The Minpp1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Multiple Inositol Polyphosphate Phosphatase 1 Protein (31-487Aa). Applications: ELISA, WB, IHC. Target Synonyms: MINPP1. Accession Number: Q9UNW1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9528 Supplier Biomatik Corporation Supplier No. CAC09946
Minpp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MINPP1. The Minpp1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Multiple Inositol Polyphosphate Phosphatase 1 Protein (31-487Aa). Applications: ELISA, WB, IHC. Target Synonyms: MINPP1. Accession Number: Q9UNW1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.