Learn More
Biomatik Corporation FBXO3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509530
Encompass_Preferred
The FBXO3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human F-box only protein 3 protein (276-376AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:200-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: F-box only protein 3, FBXO3, FBX3. UniProt: Q9UK99. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9530 Supplier Biomatik Corporation Supplier No. CAC09948
The FBXO3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant Human F-box only protein 3 protein (276-376AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:200-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: F-box only protein 3, FBXO3, FBX3. UniProt: Q9UK99. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.