Learn More
Biomatik Corporation TRAPPC2L anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509539
Encompass_Preferred
Trappc2L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRAPPC2L. The Trappc2L Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Trafficking Protein Particle Complex Subunit 2-Like Protein (1-139Aa). Applications: ELISA, WB, IHC. Target Synonyms: TRAPPC2L. Accession Number: Q9UL33. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9539 Supplier Biomatik Corporation Supplier No. CAC09957
Trappc2L Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRAPPC2L. The Trappc2L Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Trafficking Protein Particle Complex Subunit 2-Like Protein (1-139Aa). Applications: ELISA, WB, IHC. Target Synonyms: TRAPPC2L. Accession Number: Q9UL33. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.