Learn More
Biomatik Corporation PLLP anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509546
Encompass_Preferred
Pllp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLLP. The Pllp Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Plasmolipin Protein (1-35Aa). Applications: ELISA, WB, IF. Target Synonyms: PLLP. Accession Number: Q9Y342. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9546 Supplier Biomatik Corporation Supplier No. CAC09964
Pllp Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLLP. The Pllp Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Plasmolipin Protein (1-35Aa). Applications: ELISA, WB, IF. Target Synonyms: PLLP. Accession Number: Q9Y342. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.