Learn More
Biomatik Corporation MORC2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC09966
Encompass_Preferred
Morc2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MORC2. The Morc2 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Morc Family Cw-Type Zinc Finger Protein 2 Protein (2-300Aa). Applications: ELISA, WB, IHC. Target Synonyms: MORC2. Accession Number: Q9Y6X9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9548