Learn More
Biomatik Corporation NOP58 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509549
Encompass_Preferred
Nop58 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NOP58. The Nop58 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nucleolar Protein 58 Protein (1-529Aa). Applications: ELISA, WB, IF. Target Synonyms: NOP58. Accession Number: Q9Y2X3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9549 Supplier Biomatik Corporation Supplier No. CAC09967
Nop58 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NOP58. The Nop58 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nucleolar Protein 58 Protein (1-529Aa). Applications: ELISA, WB, IF. Target Synonyms: NOP58. Accession Number: Q9Y2X3. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.