Learn More
Biomatik Corporation FARP1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501509574
Encompass_Preferred
The FARP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human FERM, RhoGEF and pleckstrin domain-containing protein 1 protein (1-300AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: FERM, RhoGEF and pleckstrin domain-containing protein 1, Chondrocyte-derived ezrin-like protein, Pleckstrin homology domain-containing family C member 2, PH domain-containing family C member 2, FARP1, CDEP, PLEKHC2. UniProt: Q9Y4F1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9574 Supplier Biomatik Corporation Supplier No. CAC09992
The FARP1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human FERM, RhoGEF and pleckstrin domain-containing protein 1 protein (1-300AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB:1:500-5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: FERM, RhoGEF and pleckstrin domain-containing protein 1, Chondrocyte-derived ezrin-like protein, Pleckstrin homology domain-containing family C member 2, PH domain-containing family C member 2, FARP1, CDEP, PLEKHC2. UniProt: Q9Y4F1. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.