Learn More
Biomatik Corporation AP4B1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC10000
Encompass_Preferred
The AP4B1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human AP-4 complex subunit beta-1 protein (201-500AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-2000, IHC:1:20-1:200. Alternative Names: AP-4 complex subunit beta-1, AP4B1, AP-4 adaptor complex subunit beta, Adaptor-related protein complex 4 subunit beta-1, Beta subunit of AP-4, Beta4-adaptin. UniProt: Q9Y6B7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-9582