Learn More
Biomatik Corporation Angiopoietin Like Protein 8 (ANGPTL8) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21004
Encompass_Preferred
The Angiopoietin Like Protein 8 (ANGPTL8) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: ANGPTL8 (Met24-Leu196). Applications: WB, IHC. Alternative Names: RIFL; TD26; C19orf80; Lipasin; Betatrophin; Hepatocellular Carcinoma-Associated Protein TD26. UniProt: Q6UXH0. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9745