Learn More
Biomatik Corporation T-Box Protein 21 (TBX21) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21198
Encompass_Preferred
T-Box Protein 21 (Tbx21) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to T-Box Protein 21 (TBX21). The T-Box Protein 21 (Tbx21) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Lys160~Arg399. Applications: WB, IHC, ICC, IP. Target Synonyms: T-Box Protein 21 (TBX21). Accession Number: Q9JKD8. Gene ID: 57765. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9750