Learn More
Biomatik Corporation Wingless Type MMTV Integration Site Family, Member 3A (WNT3A) anti-Human, Bovine, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21225
Encompass_Preferred
Wingless Type Mmtv Integration Site Family, Member 3A (Wnt3A) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Wingless Type MMTV Integration Site Family, Member 3A (WNT3A). The Wingless Type Mmtv Integration Site Family, Member 3A (Wnt3A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Phe290. Applications: WB, IHC, ICC, IP. Target Synonyms: Wingless Type MMTV Integration Site Family, Member 3A (WNT3A). Accession Number: P56704. Gene ID: 89780. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9751