Learn More
Biomatik Corporation Histone Cluster 1, H2aa (HIST1H2AA) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21248
Encompass_Preferred
The Histone Cluster 1, H2aa (HIST1H2AA) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: HIST1H2AA (Ser2-Lys131). Applications: WB, IHC. Alternative Names: H2AFR; H2A Histone Family Member R. UniProt: Q96QV6. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9752