Learn More
Biomatik Corporation T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21276
Encompass_Preferred
T-Cell Immunoreceptor With Ig And Itim Domains Protein (Tigit) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT). The T-Cell Immunoreceptor With Ig And Itim Domains Protein (Tigit) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gly28~Ser246. Applications: WB, IHC, ICC, IP. Target Synonyms: T-Cell Immunoreceptor With Ig And ITIM Domains Protein (TIGIT). Accession Number: P86176. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9754