Learn More
Biomatik Corporation Nuclear Factor, Erythroid Derived 2 Like Protein 2 (NFE2L2) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21373
Encompass_Preferred
Nuclear Factor, Erythroid Derived 2 Like Protein 2 (Nfe2L2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Nuclear Factor, Erythroid Derived 2 Like Protein 2 (NFE2L2). The Nuclear Factor, Erythroid Derived 2 Like Protein 2 (Nfe2L2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ser400~Lys603. Applications: WB, IHC, ICC, IP. Target Synonyms: Nuclear Factor, Erythroid Derived 2 Like Protein 2 (NFE2L2). Accession Number: Q16236. Gene ID: 4780. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9756