Learn More
Biomatik Corporation Microtubule Associated Protein 1 Light Chain 3 Alpha (MAP1LC3a) anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509760
Encompass_Preferred
Microtubule Associated Protein 1 Light Chain 3 Alpha (Map1Lc3A) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Microtubule Associated Protein 1 Light Chain 3 Alpha (MAP1LC3a). The Microtubule Associated Protein 1 Light Chain 3 Alpha (Map1Lc3A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Gly120. Applications: WB, IHC, ICC, IP. Target Synonyms: Microtubule Associated Protein 1 Light Chain 3 Alpha (MAP1LC3a). Accession Number: Q9H492. Gene ID: 84557. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9760 Supplier Biomatik Corporation Supplier No. CAU21421
Microtubule Associated Protein 1 Light Chain 3 Alpha (Map1Lc3A) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Microtubule Associated Protein 1 Light Chain 3 Alpha (MAP1LC3a). The Microtubule Associated Protein 1 Light Chain 3 Alpha (Map1Lc3A) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Gly120. Applications: WB, IHC, ICC, IP. Target Synonyms: Microtubule Associated Protein 1 Light Chain 3 Alpha (MAP1LC3a). Accession Number: Q9H492. Gene ID: 84557. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.