Learn More
Biomatik Corporation Interleukin 1 Family, Member 9 (IL1F9) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21439
Encompass_Preferred
Interleukin 1 Family, Member 9 (Il1F9) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Interleukin 1 Family, Member 9 (IL1F9). The Interleukin 1 Family, Member 9 (Il1F9) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: His5~Lys149. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 1 Family, Member 9 (IL1F9). Accession Number: Q8R460. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9761