Learn More
Biomatik Corporation Fibroblast Growth Factor 15 (FGF15) anti-Rat, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21509
Encompass_Preferred
The Fibroblast Growth Factor 15 (FGF15) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Rat. Immunogen: FGF15 (Gly25-Lys218). Applications: WB, IHC. UniProt: Q8VI81. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9762