Learn More
Biomatik Corporation PTEN Induced Putative Kinase 1 (PINK1) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21529
Encompass_Preferred
Pten Induced Putative Kinase 1 (Pink1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTEN Induced Putative Kinase 1 (PINK1). The Pten Induced Putative Kinase 1 (Pink1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Lys262~Leu507. Applications: WB, IHC, ICC, IP. Target Synonyms: PTEN Induced Putative Kinase 1 (PINK1). Accession Number: Q9BXM7. Gene ID: 65018. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9763