Learn More
Biomatik Corporation NLR Family, Pyrin Domain Containing Protein 3 (NLRP3) anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21608
Encompass_Preferred
Nlr Family, Pyrin Domain Containing Protein 3 (Nlrp3) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to NLR Family, Pyrin Domain Containing Protein 3 (NLRP3). The Nlr Family, Pyrin Domain Containing Protein 3 (Nlrp3) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Cys671~Trp1033. Applications: WB. Target Synonyms: NLR Family, Pyrin Domain Containing Protein 3 (NLRP3). Accession Number: T1W2H6. Gene ID: 216799. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9765