Learn More
Biomatik Corporation NANOG Homeobox Protein (NANOG) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509767
Encompass_Preferred
Nanog Homeobox Protein (Nanog) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NANOG Homeobox Protein (NANOG). The Nanog Homeobox Protein (Nanog) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Val305. Applications: WB, IHC, ICC, IP. Target Synonyms: NANOG Homeobox Protein (NANOG). Accession Number: Q9H9S0. Gene ID: 79923. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9767 Supplier Biomatik Corporation Supplier No. CAU21621
Nanog Homeobox Protein (Nanog) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NANOG Homeobox Protein (NANOG). The Nanog Homeobox Protein (Nanog) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Val305. Applications: WB, IHC, ICC, IP. Target Synonyms: NANOG Homeobox Protein (NANOG). Accession Number: Q9H9S0. Gene ID: 79923. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.