Learn More
Biomatik Corporation Beclin 1 (BECN1) anti-Human, Pig, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU21701
Encompass_Preferred
The Beclin 1 (BECN1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Pig. Immunogen: BECN1 (Met1-Thr273). Applications: WB, IHC. Alternative Names: ATG6; VPS30; Coiled-Coil,Myosin-Like BCL2 Interacting Protein; ATG6 Autophagy Related 6 Homolog. UniProt: Q14457. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9768