Learn More
Biomatik Corporation DNA Damage Inducible Transcript 3 (DDIT3) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509769
Encompass_Preferred
The DNA Damage Inducible Transcript 3 (DDIT3) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: DDIT3 (Met1-Ala169). Applications: WB, IHC. Alternative Names: CEBPZ; CHOP; CHOP10; GADD153; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; Growth arrest and DNA damage-inducible GADD153. UniProt: P35638. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9769 Supplier Biomatik Corporation Supplier No. CAU21758
The DNA Damage Inducible Transcript 3 (DDIT3) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: DDIT3 (Met1-Ala169). Applications: WB, IHC. Alternative Names: CEBPZ; CHOP; CHOP10; GADD153; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; Growth arrest and DNA damage-inducible GADD153. UniProt: P35638. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.