Learn More
Biomatik Corporation Paired Box Gene 6 (PAX6) anti-Rat, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509777
Encompass_Preferred
Paired Box Gene 6 (Pax6) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Paired Box Gene 6 (PAX6). The Paired Box Gene 6 (Pax6) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Met1~Leu130. Applications: WB, IHC, ICC, IP. Target Synonyms: Paired Box Gene 6 (PAX6). Accession Number: P63016. Gene ID: 25509. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9777 Supplier Biomatik Corporation Supplier No. CAU21963
Paired Box Gene 6 (Pax6) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Paired Box Gene 6 (PAX6). The Paired Box Gene 6 (Pax6) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Met1~Leu130. Applications: WB, IHC, ICC, IP. Target Synonyms: Paired Box Gene 6 (PAX6). Accession Number: P63016. Gene ID: 25509. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.