Learn More
Biomatik Corporation Sulfatase 1 (SULF1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509781
Encompass_Preferred
Sulfatase 1 (Sulf1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sulfatase 1 (SULF1). The Sulfatase 1 (Sulf1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro609~Gly871. Applications: WB, IHC, ICC, IP. Target Synonyms: Sulfatase 1 (SULF1). Accession Number: Q8IWU6. Gene ID: 23213. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9781 Supplier Biomatik Corporation Supplier No. CAU22060
Sulfatase 1 (Sulf1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sulfatase 1 (SULF1). The Sulfatase 1 (Sulf1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro609~Gly871. Applications: WB, IHC, ICC, IP. Target Synonyms: Sulfatase 1 (SULF1). Accession Number: Q8IWU6. Gene ID: 23213. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.