Learn More
Biomatik Corporation GA Binding Protein Transcription Factor Alpha (GABPa) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU22172
Encompass_Preferred
The GA Binding Protein Transcription Factor Alpha (GABPa) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: GABPa (Ala168-Thr314). Applications: WB, IHC. Alternative Names: E4TF1-60; E4TF1A; NFT2; NRF2; NRF2A; Nuclear Respiratory Factor-2 Subunit Alpha; Transcription factor E4TF1-60. UniProt: Q06546. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9785