Learn More
Biomatik Corporation Chymase 1, Mast Cell (CMA1) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509788
Encompass_Preferred
The Chymase 1, Mast Cell (CMA1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: CMA1 (Leu9-Asn247). Applications: WB, IHC. Alternative Names: CYH; CYM; Alpha-chymase; Mast cell protease I. UniProt: P21844. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9788 Supplier Biomatik Corporation Supplier No. CAU22218
The Chymase 1, Mast Cell (CMA1) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: CMA1 (Leu9-Asn247). Applications: WB, IHC. Alternative Names: CYH; CYM; Alpha-chymase; Mast cell protease I. UniProt: P21844. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.