Learn More
Biomatik Corporation ATPase, H+/K+ Exchanging Alpha Polypeptide (ATP4a) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509791
Encompass_Preferred
The ATPase, H+/K+ Exchanging Alpha Polypeptide (ATP4a) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: ATP4a (Pro786-Cys1014). Applications: WB, IHC. Alternative Names: Gastric H,K-ATPase Alpha Subunit; H (+)-K (+)-ATPase Alpha Subunit; Proton Pump; Potassium-transporting ATPase alpha chain 1. UniProt: Q91WH7. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.
Catalog No. 50-150-9791 Supplier Biomatik Corporation Supplier No. CAU22266
The ATPase, H+/K+ Exchanging Alpha Polypeptide (ATP4a) Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse. Immunogen: ATP4a (Pro786-Cys1014). Applications: WB, IHC. Alternative Names: Gastric H,K-ATPase Alpha Subunit; H (+)-K (+)-ATPase Alpha Subunit; Proton Pump; Potassium-transporting ATPase alpha chain 1. UniProt: Q91WH7. Concentration: 500 ug/ml. Storage: -20°C. Restriction: For Research Use Only.