Learn More
Biomatik Corporation Multimerin 2 (MMRN2) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509792
Encompass_Preferred
Multimerin 2 (Mmrn2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Multimerin 2 (MMRN2). The Multimerin 2 (Mmrn2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gln23~Ala188. Applications: WB, IHC, ICC, IP. Target Synonyms: Multimerin 2 (MMRN2). Accession Number: A6H6E2. Gene ID: 105450. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9792 Supplier Biomatik Corporation Supplier No. CAU22438
Multimerin 2 (Mmrn2) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Multimerin 2 (MMRN2). The Multimerin 2 (Mmrn2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Gln23~Ala188. Applications: WB, IHC, ICC, IP. Target Synonyms: Multimerin 2 (MMRN2). Accession Number: A6H6E2. Gene ID: 105450. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.