Learn More
Biomatik Corporation Sirtuin 6 (SIRT6) anti-Rat, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509796
Encompass_Preferred
Sirtuin 6 (Sirt6) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Sirtuin 6 (SIRT6). The Sirtuin 6 (Sirt6) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Asn88~Pro288. Applications: WB, IHC, ICC, IP. Target Synonyms: Sirtuin 6 (SIRT6). Accession Number: Q4FZY2. Gene ID: 299638. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9796 Supplier Biomatik Corporation Supplier No. CAU22614
Sirtuin 6 (Sirt6) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Sirtuin 6 (SIRT6). The Sirtuin 6 (Sirt6) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Asn88~Pro288. Applications: WB, IHC, ICC, IP. Target Synonyms: Sirtuin 6 (SIRT6). Accession Number: Q4FZY2. Gene ID: 299638. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.