Learn More
Biomatik Corporation Sirtuin 1 (SIRT1) anti-Human, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509798
Encompass_Preferred
Sirtuin 1 (Sirt1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sirtuin 1 (SIRT1). The Sirtuin 1 (Sirt1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Glu244~Ala498. Applications: WB, IHC, ICC, IP. Target Synonyms: Sirtuin 1 (SIRT1). Accession Number: Q96EB6. Gene ID: 23411. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9798 Supplier Biomatik Corporation Supplier No. CAU22628
Sirtuin 1 (Sirt1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Sirtuin 1 (SIRT1). The Sirtuin 1 (Sirt1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Glu244~Ala498. Applications: WB, IHC, ICC, IP. Target Synonyms: Sirtuin 1 (SIRT1). Accession Number: Q96EB6. Gene ID: 23411. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.