Learn More
Biomatik Corporation Solute Carrier Family 30 Member 8 (SLC30A8) anti-Human, Rabbit Polyclonal Antibody, 100UG
Supplier:  Biomatik Corporation CAU22826
Encompass_Preferred
Solute Carrier Family 30 Member 8 (Slc30A8) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Solute Carrier Family 30 Member 8 (SLC30A8). The Solute Carrier Family 30 Member 8 (Slc30A8) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Thr263~Asp369. Applications: WB. Target Synonyms: Solute Carrier Family 30 Member 8 (SLC30A8). Accession Number: Q8IWU4. Gene ID: 169026. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9800