Learn More
Biomatik Corporation Pregnane X Receptor (PXR) anti-Mouse, Rabbit Polyclonal Antibody, 100UG
Catalog No. 501509802
Encompass_Preferred
Pregnane X Receptor (Pxr) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Pregnane X Receptor (PXR). The Pregnane X Receptor (Pxr) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Tyr205~Gly390. Applications: WB, IHC, ICC, IP. Target Synonyms: Pregnane X Receptor (PXR). Accession Number: O54915. Gene ID: 18171. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-9802 Supplier Biomatik Corporation Supplier No. CAU22964
Pregnane X Receptor (Pxr) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Pregnane X Receptor (PXR). The Pregnane X Receptor (Pxr) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Tyr205~Gly390. Applications: WB, IHC, ICC, IP. Target Synonyms: Pregnane X Receptor (PXR). Accession Number: O54915. Gene ID: 18171. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.